1,2,3,4,5-Pentafluoro-6-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific

PC520769-5G
APOSPC520769-5GEA 2100 EUR
APOSPC520769-5G
1,2,3,4,5-Pentafluoro-6-(trifluoromethoxy)benzene 95%
1,2,3,4,5-Pentafluoro-6-(trifluoromethoxy)benzene
Formel: C₇F₈O
Molecular Weight: 252.062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22580777
CAS-Nummer: 6161-54-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR