1-Chloro-2-fluoro-3-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific

PC520771-1G
APOSPC520771-1GEA 570 EUR
APOSPC520771-1G
1-Chloro-2-fluoro-3-(trifluoromethoxy)benzene 95%
1-Chloro-2-fluoro-3-(trifluoromethoxy)benzene
Formel: C₇H₃ClF₄O
Molecular Weight: 214,545 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22580883
CAS-Nummer: 1404194-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR