5-Bromo-1-chloro-3-fluoro-2-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific

PC520772-5G
APOSPC520772-5GEA 2300 EUR
APOSPC520772-5G
5-Bromo-1-chloro-3-fluoro-2-(trifluoromethoxy)benzene 95%
5-Bromo-1-chloro-3-fluoro-2-(trifluoromethoxy)benzene
Formel: C₇H₂BrClF₄O
Molecular Weight: 293.441 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22580729
CAS-Nummer: 1417566-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR