2,3-Difluoro-1-methyl-4-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific

PC520773-1G
APOSPC520773-1GEA 1050 EUR
APOSPC520773-1G
2,3-Difluoro-1-methyl-4-(trifluoromethoxy)benzene 95%
2,3-Difluoro-1-methyl-4-(trifluoromethoxy)benzene
Formel: C₈H₅F₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD22580955
CAS-Nummer: 1404194-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR