3,5-Dimethylbenzenesulphonyl fluoride 95%

Lieferant: Apollo Scientific

PC520785-10G
APOSPC520785-10GEA 815 EUR
APOSPC520785-10G
3,5-Dimethylbenzenesulphonyl fluoride 95%
3,5-Dimethylbenzenesulphonyl fluoride
Formel: C₈H₉FO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28987335
CAS-Nummer: 86146-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR