N-(2-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide 95%
Lieferant: Apollo Scientific
PC520787-1G
APOSPC520787-1GEA
118
EUR
APOSPC520787-1G
N-(2-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide 95%
N-(2-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide
Formel:
C₇H₅ClF₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD29067716 CAS-Nummer: 23384-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...