2-Chloro-3,6-difluorobenzotrifluoride 95%
Lieferant: Apollo Scientific
PC520794-5G
APOSPC520794-5GEA
873
EUR
APOSPC520794-5G
2-Chloro-3,6-difluorobenzotrifluoride 95%
2-Chloro-3,6-difluorobenzotrifluoride
Formel:
C₇H₂ClF₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226658 CAS-Nummer: 1099597-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...