1-Isocyanato-2-nitro-4-(trifluoromethyl)benzene

Lieferant: Apollo Scientific

PC520797-5G
APOSPC520797-5GEA 989 EUR
APOSPC520797-5G
1-Isocyanato-2-nitro-4-(trifluoromethyl)benzene
1-Isocyanato-2-nitro-4-(trifluoromethyl)benzene

May form small amount of sediment

Formel: C₈H₃F₃N₂O₃
Molecular Weight: 232,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701596
CAS-Nummer: 16588-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR