2-Fluoro-4-methoxypyridine 97%

Lieferant: Apollo Scientific

PC520798-5G
APOSPC520798-5GEA 277 EUR
APOSPC520798-5G
2-Fluoro-4-methoxypyridine 97%
2-Fluoro-4-methoxypyridine
Formel: C₆H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD12828014
CAS-Nummer: 175965-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR