2-(Trifluoromethyl)cyclohexanecarboxylic acid

Lieferant: Apollo Scientific

PC520811-1G
APOSPC520811-1GEA 899 EUR
APOSPC520811-1G
2-(Trifluoromethyl)cyclohexanecarboxylic acid
2-(Trifluoromethyl)cyclohexanecarboxylic acid
Formel: C₈H₁₁F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09811959
CAS-Nummer: 384-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR