Methyl 4-fluoro-3-morpholinosulphonyl-benzothiophene-2-carboxylate 95%

Lieferant: Apollo Scientific

PC520812-1G PC520812-250MG PC520812-500MG
APOSPC520812-1GEA 815 EUR
APOSPC520812-1G APOSPC520812-250MG APOSPC520812-500MG
Methyl 4-fluoro-3-morpholinosulphonyl-benzothiophene-2-carboxylate 95%
Methyl 4-fluoro-3-morpholinosulphonyl-benzothiophene-2-carboxylate
Formel: C₁₄H₁₄FNO₅S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09981086
CAS-Nummer: 946300-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR