2-Fluoro-1-nitro-4-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific

PC520818-1G PC520818-500MG
APOSPC520818-1GEA 582 EUR
APOSPC520818-1G APOSPC520818-500MG
2-Fluoro-1-nitro-4-(trifluoromethoxy)benzene 95%
2-Fluoro-1-nitro-4-(trifluoromethoxy)benzene
Formel: C₇H₃F₄NO₃
Molecular Weight: 225.097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194450
CAS-Nummer: 123572-64-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR