(2-Bromophenyl)-(2,2-difluorocyclopropyl)methanol 95%
Lieferant: Apollo Scientific
PC520848-100MG
APOSPC520848-100MGEA
451
EUR
APOSPC520848-100MG
(2-Bromophenyl)-(2,2-difluorocyclopropyl)methanol 95%
(2-Bromophenyl)-(2,2-difluorocyclopropyl)methanol
Formel:
C₁₀H₉BrF₂O Molecular Weight: 263,079 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20492607 CAS-Nummer: 1350637-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...