2-(2,2-Difluorocyclopropyl)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC520852-1G
APOSPC520852-1GEA
194
EUR
APOSPC520852-1G
2-(2,2-Difluorocyclopropyl)acetic acid
2-(2,2-Difluorocyclopropyl)acetic acid
Formel:
C₅H₆F₂O₂ Molecular Weight: 136,097 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19229279 CAS-Nummer: 123131-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...