(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2,2-Difluoro-1-methylcyclopropanecarboxylic acid

PC520853-1G
APOSPC520853-1GEA 154 EUR
APOSPC520853-1G
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid
Formel: C₅H₆F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07777163
CAS-Nummer: 128073-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR