1,1-Difluoro-5-azaspiro[2.3]hexane hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2-Difluoro-5-azaspiro[2.3]hexane hydrochloride

PC520858-250MG PC520858-100MG PC520858-1G
APOSPC520858-250MGEA 415 EUR
APOSPC520858-250MG APOSPC520858-100MG APOSPC520858-1G
1,1-Difluoro-5-azaspiro[2.3]hexane hydrochloride 97%
1,1-Difluoro-5-azaspiro[2.3]hexane hydrochloride
Formel: C₅H₇F₂N·ClH
Molecular Weight: 155.574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27987105
CAS-Nummer: 1630906-91-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR