Ethyl 3,3-difluorocyclobutanecarboxylate 97%
Lieferant: Apollo Scientific
PC520864-1G
APOSPC520864-1GEA
88.6
EUR
APOSPC520864-1G
Ethyl 3,3-difluorocyclobutanecarboxylate 97%
Ethyl 3,3-difluorocyclobutanecarboxylate
Formel:
C₇H₁₀F₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16660028 CAS-Nummer: 681128-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...