tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (3,3-difluoro-1-(hydroxymethyl)cyclobutyl)carbamate

PC520867-250MG
APOSPC520867-250MGEA 150 EUR
APOSPC520867-250MG
tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate
tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate
Formel: C₁₀H₁₇F₂NO₃
Molecular Weight: 237,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22566207
CAS-Nummer: 1232365-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR