4-(Trifluoromethyl)cyclohexylamine 97%

Lieferant: Apollo Scientific

PC520880-1G
APOSPC520880-1GEA 134 EUR
APOSPC520880-1G
4-(Trifluoromethyl)cyclohexylamine 97%
4-(Trifluoromethyl)cyclohexylamine
Formel: C₇H₁₂F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD09040573
CAS-Nummer: 58665-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR