3,6-Bis(trifluoromethyl)-2-chloropyridine 98%
Lieferant: Apollo Scientific
PC520889-1G
APOSPC520889-1GEA
282
EUR
APOSPC520889-1G
3,6-Bis(trifluoromethyl)-2-chloropyridine 98%
3,6-Bis(trifluoromethyl)-2-chloropyridine
Formel:
C₇H₂ClF₆N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128907 CAS-Nummer: 175136-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...