5-Trifluoromethyl-2H-pyridazin-3-one 95%

Lieferant: Apollo Scientific

Synonyme: 5-(Trifluoromethyl)pyridazin-3(2H)-one

PC520894-250MG
APOSPC520894-250MGEA 78.3 EUR
APOSPC520894-250MG
5-Trifluoromethyl-2H-pyridazin-3-one 95%
5-Trifluoromethyl-2H-pyridazin-3-one
Formel: C₅H₃F₃N₂O
Molecular Weight: 164,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09841380
CAS-Nummer: 244268-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR