1-Bromo-5-(difluoromethyl)-2-fluorobenzene 97%

Lieferant: Apollo Scientific

PC520902-25G
APOSPC520902-25GEA 0 EUR
APOSPC520902-25G
1-Bromo-5-(difluoromethyl)-2-fluorobenzene 97%
1-Bromo-5-(difluoromethyl)-2-fluorobenzene
Formel: C₇H₄BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08276255
CAS-Nummer: 886509-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR