(R)-2-(Trifluoromethyl)piperidine 97%
Lieferant: Apollo Scientific
PC520905-25G
APOSPC520905-25GEA
1740
EUR
APOSPC520905-25G
(R)-2-(Trifluoromethyl)piperidine 97%
(R)-2-(Trifluoromethyl)piperidine
Formel:
C₆H₁₀F₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226598 CAS-Nummer: 1212734-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...