6-(Trifluoromethyl)pyridine-2-carbonyl chloride 97%
Lieferant: Apollo Scientific
PC520906-1G
APOSPC520906-1GEA
223
EUR
APOSPC520906-1G
6-(Trifluoromethyl)pyridine-2-carbonyl chloride 97%
6-(Trifluoromethyl)pyridine-2-carbonyl chloride
Formel:
C₇H₃ClF₃NO Molecular Weight: 209.56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226641 CAS-Nummer: 1099597-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...