2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 95%

Lieferant: Apollo Scientific

PC520909-1G PC520909-5G
APOSPC520909-1GEA 97.3 EUR
APOSPC520909-1G APOSPC520909-5G
2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 95%
2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine
Formel: C₇H₈F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09834983
CAS-Nummer: 126069-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR