1H,1H,2H,2H-Perfluorododecanesulphonic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Henicosafluorododecane-1-sulphonic acid

PC52093-250MG
APOSPC52093-250MGEA 354 EUR
APOSPC52093-250MG
1H,1H,2H,2H-Perfluorododecanesulphonic acid
1H,1H,2H,2H-Perfluorododecanesulphonic acid
Formel: C₁₂H₅F₂₁O₃S
Molecular Weight: 628.198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334503
CAS-Nummer: 120226-60-0
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR