4-(4-Fluorobenzyl)piperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC52097-1G PC52097-25G PC52097-5G
APOSPC52097-1GEA 139 EUR
APOSPC52097-1G APOSPC52097-25G APOSPC52097-5G
4-(4-Fluorobenzyl)piperidine hydrochloride
4-(4-Fluorobenzyl)piperidine hydrochloride
Formel: C₁₂H₁₆FN·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD03840140
CAS-Nummer: 92822-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR