Ethyl 2,2-difluoro-3-methyl-butanoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2,2-difluoro-3-methyl-butyrate
PC52101-1G
APOSPC52101-1GEA
1450
EUR
APOSPC52101-1G
Ethyl 2,2-difluoro-3-methyl-butanoate
Ethyl 2,2-difluoro-3-methyl-butanoate
Formel:
C₇H₁₂F₂O₂ Molecular Weight: 166,166 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11007689 CAS-Nummer: 165544-22-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...