4-Cyano-2,6-difluorobenzoic acid 95%

Lieferant: Apollo Scientific

PC52109-100MG
APOSPC52109-100MGEA 98.6 EUR
APOSPC52109-100MG
4-Cyano-2,6-difluorobenzoic acid 95%
4-Cyano-2,6-difluorobenzoic acid
Formel: C₈H₃F₂NO₂
Molecular Weight: 183,112 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09835199
CAS-Nummer: 181073-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR