3,3,3-Trifluoro-DL-alanine methyl ester hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: Methyl 2-amino-3,3,3-trifluoropropanoate HCl, Methyl 2-amino-3,3,3-trifluoropropanoate hydrochloride, Methyl 3,3,3-trifluoro-DL-alaninate hydrochloride

PC5211-250MG
APOSPC5211-250MGEA 31.9 EUR
APOSPC5211-250MG
3,3,3-Trifluoro-DL-alanine methyl ester hydrochloride
3,3,3-Trifluoro-DL-alanine methyl ester hydrochloride
Formel: C₄H₆F₃NO₂·ClH
Molecular Weight: 193,552 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08532471
CAS-Nummer: 134297-36-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR