4-(Pentafluoroethyl)-3-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC52127-250MG
APOSPC52127-250MGEA 290 EUR
APOSPC52127-250MG
4-(Pentafluoroethyl)-3-(trifluoromethyl)benzoic acid
4-(Pentafluoroethyl)-3-(trifluoromethyl)benzoic acid
Formel: C₁₀H₄F₈O₂
Molecular Weight: 308,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381983
CAS-Nummer: 1208080-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR