2-Bromo-5-iodobenzal fluoride

Lieferant: Apollo Scientific
Achtung

PC52189-1G PC52189-250MG PC52189-5G
APOSPC52189-1GEA 575 EUR
APOSPC52189-1G APOSPC52189-250MG APOSPC52189-5G
2-Bromo-5-iodobenzal fluoride
2-Bromo-5-iodobenzal fluoride
Formel: C₇H₄BrF₂I
Lagertemperatur: Raumtemperatur
MDL: MFCD18391113
CAS-Nummer: 1261496-16-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR