(2S)-2-Amino-2-[4-chloro-3-(trifluoromethyl)phenyl]ethanol hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-2-Amino-2-(4-chloro-3-(trifluoromethyl)phenyl)ethanol hydrochloride
PC52194-250MG
APOSPC52194-250MGEA
71.2
EUR
APOSPC52194-250MG
(2S)-2-Amino-2-[4-chloro-3-(trifluoromethyl)phenyl]ethanol hydrochloride
(2S)-2-Amino-2-[4-chloro-3-(trifluoromethyl)phenyl]ethanol hydrochloride
Formel:
C₉H₉ClF₃NO·ClH Molecular Weight: 276,083 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12758208 CAS-Nummer: 1810074-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...