(2R)-2-Amino-2-[4-chloro-3-(trifluoromethyl)phenyl]ethanol hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-2-Amino-2-(4-chloro-3-(trifluoromethyl)phenyl)ethanol hydrochloride

PC52195-250MG
APOSPC52195-250MGEA 279 EUR
APOSPC52195-250MG
(2R)-2-Amino-2-[4-chloro-3-(trifluoromethyl)phenyl]ethanol hydrochloride
(2R)-2-Amino-2-[4-chloro-3-(trifluoromethyl)phenyl]ethanol hydrochloride
Formel: C₉H₉ClF₃NO·ClH
Molecular Weight: 276,083 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12758209
CAS-Nummer: 2061996-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR