Methyl-4-fluor-3-hydroxybenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-fluoro-3-hydroxybenzoate

PC5221-1G
APOSPC5221-1GEA 17.6 EUR
APOSPC5221-1G
Methyl-4-fluor-3-hydroxybenzoat
Methyl-4-fluor-3-hydroxybenzoat
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
Schmelzpunkt: 89…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797457
CAS-Nummer: 214822-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR