Methyl 4-(pentafluoroethyl)-3-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC52213-1G PC52213-250MG
APOSPC52213-1GEA 0 EUR
APOSPC52213-1G APOSPC52213-250MG
Methyl 4-(pentafluoroethyl)-3-(trifluoromethyl)benzoate
Methyl 4-(pentafluoroethyl)-3-(trifluoromethyl)benzoate
Formel: C₁₁H₆F₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD30062865
CAS-Nummer: 1980063-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR