4-(Difluoromethyl)benzylamine

Lieferant: Apollo Scientific
Achtung

PC52214-1G PC52214-250MG
APOSPC52214-1GEA 0 EUR
APOSPC52214-1G APOSPC52214-250MG
4-(Difluoromethyl)benzylamine
4-(Difluoromethyl)benzylamine
Formel: C₈H₉F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD18791602
CAS-Nummer: 754920-30-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR