2-Amino-5,6-difluorobenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Amino-2,3-difluorobenzoic acid
PC52215-250MG
APOSPC52215-250MGEA
328
EUR
APOSPC52215-250MG
2-Amino-5,6-difluorobenzoic acid
2-Amino-5,6-difluorobenzoic acid
Formel:
C₇H₅F₂NO₂ Molecular Weight: 173,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06808540 CAS-Nummer: 442134-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...