2-Amino-3,6-difluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC52216-250MG
APOSPC52216-250MGEA 191 EUR
APOSPC52216-250MG
2-Amino-3,6-difluorobenzoic acid
2-Amino-3,6-difluorobenzoic acid
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569542
CAS-Nummer: 825654-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR