2-Amino-5-chloro-4-fluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC52217-1G PC52217-250MG
APOSPC52217-1GEA 422 EUR
APOSPC52217-1G APOSPC52217-250MG
2-Amino-5-chloro-4-fluorobenzoic acid
2-Amino-5-chloro-4-fluorobenzoic acid
Formel: C₇H₅ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09861352
CAS-Nummer: 351367-77-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR