5,6-Difluoro-1H-1,3-benzodiazole-2-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC52218-250MG
APOSPC52218-250MGEA
545
EUR
APOSPC52218-250MG
5,6-Difluoro-1H-1,3-benzodiazole-2-carbaldehyde
5,6-Difluoro-1H-1,3-benzodiazole-2-carbaldehyde
Formel:
C₈H₄F₂N₂O Molecular Weight: 182,127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18432482 CAS-Nummer: 1263379-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...