5-Chloro-6-fluoro-1H-1,3-benzodiazole-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC52219-250MG
APOSPC52219-250MGEA 400 EUR
APOSPC52219-250MG
5-Chloro-6-fluoro-1H-1,3-benzodiazole-2-carbaldehyde
5-Chloro-6-fluoro-1H-1,3-benzodiazole-2-carbaldehyde
Formel: C₈H₄ClFN₂O
Molecular Weight: 198,582 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18432481
CAS-Nummer: 1263378-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR