2-Amino-3,4,5-trifluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC52220-250MG
APOSPC52220-250MGEA 328 EUR
APOSPC52220-250MG
2-Amino-3,4,5-trifluorobenzoic acid
2-Amino-3,4,5-trifluorobenzoic acid
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774158
CAS-Nummer: 531529-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR