1-(Trifluoroacetyl)piperidine-3-carboxylic acid 95%

Lieferant: Apollo Scientific

PC52448-250MG PC52448-500MG
APOSPC52448-250MGEA 173 EUR
APOSPC52448-250MG APOSPC52448-500MG
1-(Trifluoroacetyl)piperidine-3-carboxylic acid 95%
1-(Trifluoroacetyl)piperidine-3-carboxylic acid
Formel: C₈H₁₀F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09908273
CAS-Nummer: 1099017-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR