5-Chloro-2-fluoro-4-iodoaniline 95%

Lieferant: Apollo Scientific

PC52463-1G PC52463-250MG PC52463-5G
APOSPC52463-1GEA 543 EUR
APOSPC52463-1G APOSPC52463-250MG APOSPC52463-5G
5-Chloro-2-fluoro-4-iodoaniline 95%
5-Chloro-2-fluoro-4-iodoaniline
Formel: C₆H₄ClFIN
Molecular Weight: 271.459 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1935087-03-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR