5-Chloro-2-fluoro-4-iodoaniline 95%
Lieferant: Apollo Scientific
PC52463-1G
PC52463-250MG
PC52463-5G
APOSPC52463-1GEA
543
EUR
APOSPC52463-1G
APOSPC52463-250MG
APOSPC52463-5G
5-Chloro-2-fluoro-4-iodoaniline 95%
5-Chloro-2-fluoro-4-iodoaniline
Formel:
C₆H₄ClFIN Molecular Weight: 271.459 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1935087-03-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...