2,6-Dichlor-4-(trifluormethoxy)benzylbromid 95%

Lieferant: Apollo Scientific

PC52464-5G
APOSPC52464-5GEA 567 EUR
APOSPC52464-5G
2,6-Dichlor-4-(trifluormethoxy)benzylbromid 95%
2,6-Dichlor-4-(trifluormethoxy)benzylbromid
Formel: C₈H₄BrCl₂F₃O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 886503-08-8
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR