3-Chloro-5-(trifluoromethoxy)benzylamine 95%

Lieferant: Apollo Scientific

PC52465-1G
APOSPC52465-1GEA 614 EUR
APOSPC52465-1G
3-Chloro-5-(trifluoromethoxy)benzylamine 95%
3-Chloro-5-(trifluoromethoxy)benzylamine
Formel: C₈H₇ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18393825
CAS-Nummer: 916210-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR