2,5-Bis(trifluoromethyl)toluene 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-1,4-bis(trifluoromethyl)benzene

PC52480-10G PC52480-1G PC52480-5G
APOSPC52480-10GEA 574 EUR
APOSPC52480-10G APOSPC52480-1G APOSPC52480-5G
2,5-Bis(trifluoromethyl)toluene 98%
2,5-Bis(trifluoromethyl)toluene
Formel: C₉H₆F₆
Molecular Weight: 228.134 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2385323-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR