2-Hydroxy-5-(pentafluorothio)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC5251-100MG PC5251-1G PC5251-250MG
APOSPC5251-100MGEA 116 EUR
APOSPC5251-100MG APOSPC5251-1G APOSPC5251-250MG
2-Hydroxy-5-(pentafluorothio)benzaldehyde
2-Hydroxy-5-(pentafluorothio)benzaldehyde
Formel: C₇H₅F₅O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11501046
CAS-Nummer: 1159512-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR